Socorex™ Calibra® 822 Digital Single-Channel Micropipettors, Variable Volume, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Sleek micropipettors are ideal for gel sequencing and biotechnology applications.
- Micropipettors are also color-coded to ensure proper tip selection
- Small sample volumes can be reproducibly dispensed
- Autoclavable
They are equipped with twin cams with precalibrated steps to provide true digital volume entry, high performance, and calibration stability. Plunger assembly features fluoropolymer resin and stainless steel construction for a high degree of chemical resistance.
Ordering information: Supplied with a manufacturer’s three-year warranty on parts and labor, including two years on original calibration. Includes an Allen wrench for in-lab calibration.
Frequently Bought Together