Nalgene® Wide Mouth Centrifuge Bottles with Screw Cap, Polystyrene, Sterile, Conical-Bottom, Thermo Scientific
Catalog # 21010-610
Supplier: Thermo Fisher Scientific
Specifications
- Ø:61.5 mm
- Capacity:177 ml (6 oz.)
- Description:Sterile Centrifuge Bottle with Cap
- Length:135.6 mm
- Cat. no.:21010-610
- Maximum RCF:5800×g
- Supplier No.:3145-0175
Specifications
About this item
This sterile, graduated bottle is ideal for cell culture applications.
Note: Must be used with Nalgene® adapter for conical-bottom centrifuge bottles, listed below. Nalgene® labware is for laboratory use only, not for in vitro diagnosis or parenterals. For proper performance, fill bottles to at least 80% of the total capacity.
It is suitable for spinning at low speeds. The wide mouth allows easier filling and decanting. Cap and bottle are gamma radiation sterilized. Leakproof.
Ordering information: Supplied sterile, individually wrapped, with sterile polyethylene screw cap. Sealing cap is not supplied sterile.