Socorex™ Acura® Single-Channel Pipettors, Variable Volume, DWK Life Sciences
Lightweight pipettors are designed for the accurate, precise pipetting of solutions, reagents, acids, serums, and other liquids, and for gel-sequencing applications.
- Easy swift-set calibration requires no additional tools beyond a balance and distilled water
- Adjust volume setting from high to low in one click
- Setting wheel locks eliminate accidental volume alteration
- Dispensing is accomplished with a single plunger stroke
- Autoclavable to 121 °C (250 °F) without disassembly or recalibration
A differentiated overshoot function and separate push buttons for dispensing and tip ejection reduce dead volume and prevent accidental de-tipping. The glove-friendly JustTip™ adjustable tip ejector allows the use of most pipette tips. The stainless steel and PVDF plunger assembly is chemically resistant.
: Supplied with a three-year manufacturer’s warranty, including a two-year warranty on the original calibration. Pipettors 83009-172 and -174 feature cellulose paper filters that may be used with polypropylene tips to prevent cross-contamination. Color-coded button caps are available (see 83009-204 series).
Frequently Bought Together