Biohit Family mLINE® Single-Channel Mechanical Pipettors, Variable Volume, Sartorius | Avantor
Biohit Family mLINE® Single-Channel Mechanical Pipettors, Variable Volume, Sartorius
Biohit <i>Family</i> mLINE® Single-Channel Mechanical Pipettors, Variable Volume, Sartorius
Supplier:  Sartorius
Certificates
Biohit <i>Family</i> mLINE® Single-Channel Mechanical Pipettors, Variable Volume, Sartorius
Biohit Family mLINE® Single-Channel Mechanical Pipettors, Variable Volume, Sartorius
8 Options Available Below
Product Details & Documents
Sartorius mLINE® Mechanical Pipettors are an advanced and ergonomic choice for quality-conscious users who perform long and repetitive manual pipetting. Designed for repetitive and long-lasting procedures in order to prevent work-related hand, arm, and shoulder disorders or Repetitive Strain Injury (RSI).
  • Excellent ergonomics to protect from injuries with exceptionally light pipetting and tip ejection forces
  • High accuracy and precision, including in repetitive, long-lasting procedures
  • Minimized risk of contamination with Safe-Cone Filters and full autoclavability
  • Increased safety with volume lock preventing accidental volume changes while pipetting
Sturdy yet lightweight pipettors meet the strictest performance, ergonomic, and safety criteria.
Pipettors feature a clear display with large, easy-to-read numbers. Counter locking mechanism prevents accidental volume changes during pipetting. Volume range color coding, ID tag, and color coding caps allow easy pipettor identification.
Simple to clean and maintain with only three parts to disassemble. Fully autoclavable.
Unique loading mechanism allows tip to be loaded with constant force. This ensures the tip is correctly sealed and can be ejected with minimum force. Thermal insulation of internal components provides improved accuracy. The fast, light, stepless volume adjustment and small increments allow optimal precision.
Informations sur les accessoires: Linear stand accessory is suitable for all Sartorius mechanical and electronic pipettors.
Documents
Product Family Options
Variable Volume Single Channel Mechanical Pipettors
Informations sur le produit
  • Volume
    Increments
    Accuracy
    Precision
    Availability
    Price
  • 0.3 - 3 µl
    0.002 µl
    ±10.0% at 0.3 µl, ±1.3% at 3 µl
    ±6.0% at 0.3 µL, ±0.8% at 3 µL
    Discontinued
    Supplier Number: 725010
    Spécifications:
    More
    Specifications
    Cat. No.
    14005-976
  • 0.1 - 3 µl
    0.002 µl
    ±10.0% at 0.3 µl, ±1.3% at 3 µl
    ±6.0% at 0.3 µL, ±0.8% at 3 µL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $678.90
    $678.90
    Each
       $678.90
    Supplier Number: 725010
    Spécifications:
    More
    Specifications
    Cat. No.
    CA14005-976
  • 1000 - 10000 µl
    20 µl
    ±3.0% at 1000 µl, ±0.6% at 10,000 µl
    ±0.6% at 1000 µL, ±0.2% at 10,000 µL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $678.90
    $678.90
    Each
       $678.90
    Supplier Number: 725090
    Spécifications:
    More
    Specifications
    Cat. No.
    CA89079-110
  • 0.5 - 10 µl
    0.01 µl
    ±2.5% at 1 µl, ±1.0% at 10 µl
    ±1.5% at 1 µL, ±0.6% at 10 µL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $678.90
    $678.90
    Each
       $678.90
    Supplier Number: 725020
    Spécifications:
    More
    Specifications
    Cat. No.
    CA47745-544
  • 2 - 20 µl
    0.02 µl
    ±3.0% at 2 µl, ±0.9% at 20 µl
    ±2.0% at 2 µL, ±0.4% at 20 µL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $678.90
    $678.90
    Each
       $678.90
    Supplier Number: 725030
    Spécifications:
    More
    Specifications
    Cat. No.
    CA47745-545
  • 10 - 100 µl
    0.1 µl
    ±2.0% at 10 µl, ±0.8% at 100 µl
    ±1.0% at 10 µL, ±0.15% at 100 µL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $678.90
    $678.90
    Each
       $678.90
    Supplier Number: 725050
    Spécifications:
    More
    Specifications
    Cat. No.
    CA47745-546
  • 20 - 200 µl
    0.2 µl
    ±2.3% at 20 µl, ±0.6% at 200 µl
    ±0.9% at 20 µL, ±0.15% at 200 µL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $678.90
    $678.90
    Each
       $678.90
    Supplier Number: 725060
    Spécifications:
    More
    Specifications
    Cat. No.
    CA47745-548
  • 100 - 1000 µl
    1 µl
    ±2.0% at 100 µl, ±0.7% at 1000 µl
    ±0.5% at 100 µL, ±0.2% at 1000 µL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $678.90
    $678.90
    Each
       $678.90
    Supplier Number: 725070
    Spécifications:
    More
    Specifications
    Cat. No.
    CA47745-550
  • 500 - 5000 µl
    10 µl
    ±2.0% at 500 µl, ±0.5% at 5000 µl
    ±0.6% at 500 µL, ±0.2% at 5000 µL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $678.90
    $678.90
    Each
       $678.90
    Supplier Number: 725080
    Spécifications:
    More
    Specifications
    Cat. No.
    CA47745-552
  • 1 - 10 µl
    0.01 µl
    ±2.5% at 1 µl, ±1.0% at 10 µl
    ±1.5% at 1 µL, ±0.6% at 10 µL
    Discontinued
    Supplier Number: 725020
    Spécifications:
    More
    Specifications
    Cat. No.
    47745-544
  • 2 - 20 µl
    0.02 µl
    ±3.0% at 2 µl, ±0.9% at 20 µl
    ±2.0% at 2 µL, ±0.4% at 20 µL
    Discontinued
    Supplier Number: 725030
    Spécifications:
    More
    Specifications
    Cat. No.
    47745-545
  • 10 - 100 µl
    0.1 µl
    ±2.0% at 10 µl, ±0.8% at 100 µl
    ±1.0% at 10 µL, ±0.15% at 100 µL
    Discontinued
    Supplier Number: 725050
    Spécifications:
    More
    Specifications
    Cat. No.
    47745-546
  • 20 - 200 µl
    0.2 µl
    ±2.3% at 20 µl, ±0.6% at 200 µl
    ±0.9% at 20 µL, ±0.15% at 200 µL
    Discontinued
    Supplier Number: 725060
    Spécifications:
    More
    Specifications
    Cat. No.
    47745-548
  • 100 - 1000 µl
    1 µl
    ±2.0% at 100 µl, ±0.7% at 1000 µl
    ±0.5% at 100 µL, ±0.2% at 1000 µL
    Discontinued
    Supplier Number: 725070
    Spécifications:
    More
    Specifications
    Cat. No.
    47745-550
  • 500 - 5000 µl
    10 µl
    ±2.0% at 500 µl, ±0.5% at 5000 µl
    ±0.6% at 500 µL, ±0.2% at 5000 µL
    Discontinued
    Supplier Number: 725080
    Spécifications:
    More
    Specifications
    Cat. No.
    47745-552
  • 1000 - 10000 µl
    20 µl
    ±3.0% at 1000 µl, ±0.6% at 10,000 µl
    ±0.6% at 1000 µL, ±0.2% at 10,000 µL
    Discontinued
    Supplier Number: 725090
    Spécifications:
    More
    Specifications
    Cat. No.
    89079-110
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Linear stand, holds up to 7 pipettors
    Linear stand, holds up to 7 pipettors
    CA80087-478
    Log in pour la disponibilité
    Each - $280.88
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Carousel stand for 6 pipettes
    Carousel stand for 6 pipettes
    75870-918
    Tacta® and mLINE® pipettors
    Log in pour la disponibilité
    Each - $437.79