ContactUsLinkComponent
Automated Electronic Sterilizers, Heidolph Tuttnauer
Automated Electronic Sterilizers, Heidolph Tuttnauer
Supplier:  Heidolph NA, LLC
Certificates
undefined
Automated Electronic Sterilizers, Heidolph Tuttnauer
Supplier:  Heidolph NA, LLC
Specifications
  • Catalog No:
  • CA89423-436
  • Max. working pressure:
  • 40 psi
  • Phase:
  • 1
  • Plug type:
  • Nema 5-15R (120V units), Nema 6-15R (220V units)
  • Pump fuse:
  • Water Pump: 1.25 amps, Air Pump: 2.0 amps (120V units)/1.25 amps (220V units)