Finnpipette® F2 Single-Channel Variable Volume Pipettors, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
Finnpipette® F2 variable volume pipettors offer outstanding accuracy and precision in laboratory based liquid dispensing procedures
- Each lightweight device offers remarkably smooth pipetting action
- Pipettors are fully autoclavable and easy to service
Pipettors are equipped with an advanced volume gearing mechanism (AVG) designed to improve accuracy and precision. An enlarged digit display improves ease of use, and color coding simplifies instrument selection.
These pipettors combine the established Finnpipette® features with state-of-the-art innovations to create a reliable and ergonomic high-performance device.
Ordering information: Finnpipette® F2 devices are supplied with a five-year manufacturer's warranty.
Frequently Bought Together