MLA Micro D-Tipper Pipettors, Fixed Volume, VistaLab
: VistaLab
MLA-style Micro D-Tipper pipettors provide dependable performance and repeatable results in applications using microvolume sample sizes.
- In-lab calibration is performed with minimal effort
- Color-coded for quick and convenient volume identification
- Tip ejection requires a simple upward lift on the bonnet
Manufactured for durable, extended use, these pipettors are made of precision engineered aluminum and stainless steel, and feature a tapered design. These easy-to-use pipettors use a smooth two-stroke action (with overblow) to ensure precise results in repetitive procedures.
Frequently Bought Together