Finnpipette™ F1 Fixed Volume Single-Channel Pipettes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Great for set protocols and QA environments, Thermo Scientific™ Finnpipette™ F1 Fixed Volume Pipettes deliver a light, smooth pipetting action and an adjustable finger rest to maximize your comfort.
- Smooth pipetting action
- Adjustable finger rest
- Lightweight design
- Five-year warranty
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI. The 120° adjustable finger rest, for both right- and left-handed operator comfort, allows the hand to relax during pipetting cycles.
A 150% increase in air boost ensures efficient delivery of micro-volumes and prevent capillary action in 50μl models and below.
Certifications: The Finnpipette F1 Pipette together with compatible Finntip tips is CE marked in accordance with IVD Directive in Europe.
Accessories information: Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
Ordering information: Five-year warranty with convenient online registration; may vary by country.
Frequently Bought Together