VWR® Low-Temperature Covers for Modular Heating Blocks
Supplier: VWR International
Jump to:
À propos de cet article
The Plexiglas® cover reduces air flow for additional temperature stability in low-temperature applications.
- Now with Best-in-Class, Exclusive 5-Year Warranty
Two sides open 6.4mm (1/4").
3 Options Available Below
Product Details & Documents
The Plexiglas® cover reduces air flow for additional temperature stability in low-temperature applications.
- Now with Best-in-Class, Exclusive 5-Year Warranty
Two sides open 6.4mm (1/4").