VWR® Modular Heating Blocks for Microcentrifuge Tubes
Supplier: VWR International
À propos de cet article
Constructed from a solid anodized aluminum block.
- Now with Best-in-Class, Exclusive 5-Year Warranty
The close contact of tubes to block walls allows for maximum heat retention. Single block size.
6 Options Available Below
Product Details & Documents
Constructed from a solid anodized aluminum block.
- Now with Best-in-Class, Exclusive 5-Year Warranty
The close contact of tubes to block walls allows for maximum heat retention. Single block size.
Specifications for Product Family
Spécifications
- Catalog No:
- 12985-048
- 13259-000
- 13259-002
- 13259-286
- 12985-050
- 12985-044
- Dimensions:
- 9.5L x 7.6W x 5.1H cm (33/4 x 3 x 2")
- 9.5L x 7.6W x 5.1H cm (33/4 x 3 x 2")
- 9.5L x 7.6W x 5.1H cm (33/4 x 3 x 2")
- 9.5L x 7.6W x 5.1H cm (33/4 x 3 x 2")
- 9.5L x 7.6W x 5.1H cm (33/4 x 3 x 2")
Spécifications
Product Family Options
Informations sur le produit
- DésignationFor Tube VolumeFor Use WithNo. of WellsWell DepthWell DiameterAvailabilityPrice
- 2 mL Microcentrifuge Tube Block2.0 mlVWR®/Eppendorf® Microcentrifuge Tubes2038.1 mm11.5 mmLog in pour la disponibilitéEach$463.01Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.12985-048Dimensions9.5L x 7.6W x 5.1H cm (33/4 x 3 x 2")Product available on GSA Advantage® - 0.5 mL Microcentrifuge Tube Block0.5 mlVWR® Tapered Microcentrifuge Tubes3027.6 mm7.9 mmLog in pour la disponibilitéEach$393.41Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.13259-000Dimensions9.5L x 7.6W x 5.1H cm (33/4 x 3 x 2")Product available on GSA Advantage® - Block, Microcentrifuge Tube1.5 mlEppendorf® Microcentrifuge Tubes2036.9 mm11.5 mmLog in pour la disponibilitéEach$421.33Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.13259-002Dimensions9.5L x 7.6W x 5.1H cm (33/4 x 3 x 2")Product available on GSA Advantage® - 1.5 mL Microcentrifuge Tube Block1.5 mlVWR® Microcentrifuge Tubes2039.1 mm11.1 mmLog in pour la disponibilitéEach$390.16Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.13259-286Dimensions9.5L x 7.6W x 5.1H cm (33/4 x 3 x 2")Product available on GSA Advantage® - Block, Microcentrifuge Tube2.0 mlCorning® Microcentrifuge Tubes2038.1 mm10.9 mmLog in pour la disponibilitéEach$418.01Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.12985-050Dimensions9.5L x 7.6W x 5.1H cm (33/4 x 3 x 2")Product available on GSA Advantage® - Block, Microcentrifuge Tube1.5 mlVWR® Microcentrifuge Tubes1039.1 mm11.1 mmLog in pour la disponibilitéEach$253.15Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.12985-044DimensionsProduct available on GSA Advantage®