VWR® Modular Heating Block for Titer Plates
Catalogue # 13259-295
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Description:Block, 96 Test Well Plate, Titer Plate
- Cat. no.:13259-295
- L×W×H:15.1 x 9.5 x 5.7 cm (6 x 33/4 x 21/4")
- Block Size:Double
Spécifications
À propos de cet article
Heating block is ideal for use with 96-well or 384-well titer plates.
- Now with Best-in-Class, Exclusive 5-Year Warranty
It features a recessed well for better stability and a flat surface appropriate for flat or round-bottom plates. Double block size fits only 2, 4, or 6 block heaters.