Axygen® brand automation tips are engineered to the exact specifications of the original equipment manufacturers’ tips, ensuring complete compatibility and a consistent fit.
- Engineered to the exact specifications to ensure compatibility and consistent fit
- Certified DNase-/RNase-free and nonpyrogenic for complete protection against contamination
- Maxymum Recovery® versions also available
- Filtered version available in most tip styles to prevent aerosol and liquid contamination
Axygen® offers a comprehensive line of tips for Beckman Coulter®, Dynamic Devices, Hamiltion Starline, and ProGroup Wellpro automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Available in nonsterile, sterile, non-filtered, filtered, wide-bore, fine point, and extended length formats. Clear polypropylene tips are available in 96-well and 384-well formats. Also available in Maxymum Recovery® to further improve accuracy and precision while reducing sample loss.
- Catalog No:
- 47734-686
- 80082-910
- 47745-460
- 47745-472
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa