Hi Guard® Full Coverage Boots
New Product
Halyard footwear product line features innovative fabrics and styling to deliver the right amount of protection for the task.
- Plastic film coating
- Traction slips
- Single use
- Elastic at the ankle
- Ultra-sonically bonded seams
- Shoe and lower leg coverage
Protective SMS fabric with film coating on foot area; adhesive traction strips. Moderate fluid contact above the shoe.
Frequently Bought Together