Labcor™ Pipettor Solution Basins, Biotix
: Avantor
Disposable basins are designed for 8- and 12-channel pipettors
- Angled sides and V-shaped bottom permit easy access to solution
- Pre-sterile basins are electron beam sterilized and RNase-, DNase-, and pyrogen-free
- Polyvinyl chloride material allows the contents to be clearly viewed
Basins are available pre-sterile or nonsterile and constructed of either polyvinyl chloride or polystyrene.
Disposable basins are designed for 8- and 12-channel pipettors
- Angled sides and V-shaped bottom permit easy access to solution
- Pre-sterile basins are electron beam sterilized and RNase-, DNase-, and pyrogen-free
- Polyvinyl chloride material allows the contents to be clearly viewed
Basins are available pre-sterile or nonsterile and constructed of either polyvinyl chloride or polystyrene.
Product Family Options
- MatièreCapacitéHauteurLongueurLargeurSterilityPacked
- Polystyrene100 ml3.2 cm (1¼")15.2 cm (6")8 cm (3⁵⁄₃₂")Non sterileBulkCase of 100$169.04
- Polystyrene100 ml3.2 cm (1¼")15.2 cm (6")8 cm (3⁵⁄₃₂")Pre-sterileIndividually Wrapped-
- Polystyrene100 ml3.2 cm (1¼")15.2 cm (6")8 cm (3⁵⁄₃₂")Pre-sterile5/Bag-
- PVC55 ml2.5 cm (1")14.3 cm (5⅝")7 cm (2¾")Non sterileBulk-
- Polystyrene55 ml2.5 cm (1")14.3 cm (5⅝")7 cm (2¾")Pre-sterileIndividually Wrapped-
- Polystyrene55 ml2.5 cm (1")14.8 cm (5²⁷⁄₃₂")6.4 cm (2½")Pre-sterile5/Bag-