Supelco® pH Buffer Solutions, MilliporeSigma
Supelco® pH Buffer Solutions, MilliporeSigma
Supelco® pH Buffer Solutions, MilliporeSigma
Supplier:  MilliporeSigma
Certificates
Supelco® pH Buffer Solutions, MilliporeSigma
Supelco® pH Buffer Solutions, MilliporeSigma
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Certified reference material, traceable to NIST Standard.
  • Ready-to-use solution available in a sealed bottle with convenient packaging to protect against contamination
Available as colorless or color-coded solutions.
Product Family Options
Product Information
  • Désignation
    pH Accuracy
    Type de conditionnement
    Taille
    pH
    Color
    Availability
    Price
  • ±0.01
    Plastic bottle
    500 ml
    1.62
    Colorless
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $54.78
    $54.78
    Each(1/Itm/Bx)
       $54.78
    Supplier Number: EM01822-500ML
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.02
    Plastic bottle
    500 ml
    2.00
    Colorless
    Discontinued
    Supplier Number: -
  • ±0.02
    Plastic bottle
    500 ml
    2.00
    Colorless
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.01
    Plastic bottle
    500 ml
    4.00
    Red
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic receptable in fibreboard box
    20 L
    4.00
    Red
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic bottle
    500 ml
    4.00
    Red
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic bottle
    500 ml
    4.00
    Clear
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic bottle
    500 ml
    4.00
    Colorless
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $49.42
    $49.42
    Each(1/Itm/Bx)
       $49.42
    Supplier Number: BX16341-500ML
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.02
    Plastic bottle
    500 ml
    5.00
    Clear
    Discontinued
    Supplier Number: -
  • ±0.02
    Plastic bottle
    500 ml
    5.00
    Colorless
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.02
    Plastic bottle
    500 ml
    6.00
    Clear
    Discontinued
    Supplier Number: -
  • ±0.02
    Plastic bottle
    500 ml
    6.00
    Colorless
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.01
    Plastic bottle
    500 ml
    6.86
    Colorless
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic bottle
    500 ml
    6.86
    Colorless
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.01
    Plastic bottle
    500 ml
    7.00
    Yellow
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic receptable in fibreboard box
    20 L
    7.00
    Yellow
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic bottle
    4 L
    7.00
    Yellow
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic bottle
    500 ml
    7.00
    Clear
    Discontinued
    Supplier Number: -
  • ±0.03
    Plastic bottle
    500 ml
    7.00
    Yellow
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.03
    Plastic receptable in fibreboard box
    20 L
    7.00
    Yellow
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.03
    Plastic bottle
    4 L
    7.00
    Yellow
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.01
    Plastic bottle
    500 ml
    7.00
    Colorless
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $50.46
    $50.46
    Each(1/Itm/Bx)
       $50.46
    Supplier Number: BX16351-500ML
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.02
    Plastic bottle
    500 ml
    8.00
    Clear
    Discontinued
    Supplier Number: -
  • ±0.02
    Plastic bottle
    500 ml
    8.00
    Colorless
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.01
    Plastic bottle
    500 ml
    10.00
    Colorless
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic bottle
    500 ml
    10.00
    Blue
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic bottle
    500 ml
    10.00
    Blue
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic receptable in fibreboard box
    20 L
    10.00
    Blue
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic bottle
    500 ml
    10.00
    Colorless
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic bottle
    500 ml
    10.00
    Blue
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic bottle
    500 ml
    10.00
    Colorless
    Discontinued
    Supplier Number: -
  • ±0.02
    Plastic bottle
    500 ml
    10.00
    Blue
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.01
    Plastic bottle
    500 ml
    10.00
    Blue
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.01
    Plastic receptable in fibreboard box
    20 L
    10.00
    Blue
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.01
    Plastic bottle
    4 L
    10.00
    Blue
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.01
    Plastic bottle
    500 ml
    10.00
    Colorless
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.02
    Plastic bottle
    500 ml
    10.00
    Colorless
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $50.46
    $50.46
    Each(1/Itm/Bx)
       $50.46
    Supplier Number: BX16421-500ML
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.01
    Plastic bottle
    500 ml
    12.45
    Colorless
    Discontinued
    Supplier Number: -
  • ±0.01
    Plastic bottle
    500 ml
    12.45
    Colorless
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • ±0.02
    3.00
    Discontinued
    Supplier Number: -
  • Discontinued
    Supplier Number: -