MASTERBLOCK® 1536-Well Deep Well Plates, Polypropylene, Greiner Bio-One
82051-410
Product available on GSA Advantage®
- Color:Clear
- Cat. no.:82051-410
- Working Well Volume:3 - 15 µl
- Maximum Well Volume:18 µL
- Supplier No.:782270
These conical-bottom microplates feature square-round internal well geometry to minimize wicking and air bubble formation.
- Free of RNase, DNase, and DNA-polymerase inhibitors
- Polypropylene construction
- Low-binding and biologically inert
- Autoclavable
Deep well microplates are manufactured of ClearChoice™ virgin resin in molds specifically engineered for polypropylene
Microplates are RNase-free, DNase-free, nonpyrogenic, and free of DNA-polymerase-inhibitors.
Produced with uniform dimensions and tolerances to meet SBS recommendations for compatiblity with equipment and robotics. Polypropylene is biologically inert, low binding, autoclavable, and resistant to many standard lab chemicals.