ContactUsLinkComponent
Neptune® Universal Pipette Tips
Neptune® Universal Pipette Tips
Supplier:  Biotix
   Sustainable Options Available
Certificates
undefined
Neptune® Universal Pipette Tips
Supplier:  Biotix
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • 89140-880
  • 89425-636
  • 89425-638
  • 89140-882
  • 89140-918
  • 10144-788
  • 89140-886
  • 89140-890
  • 89425-650
  • 89168-762
  • 89168-766
  • 89140-896
  • 89168-764
  • 89140-894
  • 89140-878
  • Environmentally Preferable:
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Environmental certification:
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%