ContactUsLinkComponent
Neptune® S³ Low Retention Pipette Tips
Neptune® S³ Low Retention Pipette Tips
Supplier:  Biotix
   Sustainable Options Available
Certificates
undefined
Neptune® S³ Low Retention Pipette Tips
Supplier:  Biotix
   Sustainable Options Available
Specifications
  • Catalog No:
  • 89140-884
  • 89140-888
  • 89140-892
  • 89425-648
  • 89168-760
  • 89140-100
  • 89168-736
  • 89168-758
  • 89140-130
  • 89140-128
  • 89140-898
  • 89140-098
  • 89168-734
  • 89140-096
  • 89168-732
  • Environmentally Preferable:
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Environmental certification:
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%