ESP™ Pipet Tip Reloading Systems, Neptune™
ESP™ Pipet Tip Reloading Systems, Neptune™
   Options durables disponibles
Supplier:   Ohaus
ESP™ Pipet Tip Reloading Systems, Neptune™
Supplier:  Ohaus
   Options durables disponibles
Certificates
ESP™ Pipet Tip Reloading Systems, Neptune™
ESP™ Pipet Tip Reloading Systems, Neptune™
15 Options Available Below
Product Details & Documents
Revolutionary ESP reload system offers the environmentally friendly reuse of existing trays.
  • Autoclavable
  • Good sustainability
  • Superior stability, firm placement
  • Maximum space saving design
Eco-friendly empty pipette tips trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%.
Unique transfer system minimizes plastic waste by as much as 90%. Reload 10 trays in 90 seconds. Integrated transfer card prevents contamination by minimizing handling. Universal pipette tips are made from virgin polypropylene. The thin wall design provides an optimal seal. Tips featuring the S³ polymer deliver to within 0.1% of indicated volume, ensuring a more accurate application.
Certifications: Certified free of RNase, DNase, pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89425-640
  • 89168-748
  • 89168-772
  • 89168-768
  • 89217-464
  • 89140-104
  • 89140-902
  • 89168-742
  • 89140-900
  • 89168-738
  • 76311-242
  • 76311-290
  • 76311-244
  • 76311-246
  • 76311-248
  • Environmentally Preferable:
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Environmental certification:
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Empty rack intended for Reload and Bulk Refills
  • FSC-STD-40-004 V3-1 and FSC-C154485; Empty rack intended for Reload and Bulk Refills
  • FSC-STD-40-004 V3-1 and FSC-C154485; Empty rack intended for Reload and Bulk Refills
  • FSC-STD-40-004 V3-1 and FSC-C154485; Empty rack intended for Reload and Bulk Refills
  • FSC-STD-40-004 V3-1 and FSC-C154485; Empty rack intended for Reload and Bulk Refills
Spécifications
Product Family Options
Polypropylene Tips
Informations sur le produit
  • Tip Style
    Volume
    Tip Color
    For Use With
    Packed
    Availability
    Price
  • ESP™ Pipette Tip Reloading Systems, Neptune™
    1000 µl
    Clear
    96/Card
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 960
    Pack of 960 - $63.77
    $63.77
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 4800
    Case of 4800 - $318.86
    $318.86
    Prix promotionnel pour les nouveaux clients
    Pack of 960
       $63.77
    Case of 4800
       $318.86
    Supplier Number: 63302170
    Spécifications:
    Product available on GSA Advantage®
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    89425-640
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
    Product available on GSA Advantage®
      Waste Reducing|Sustainable Packaging
  • Extended Length Tips
    10 µl
    Clear
    96/Card
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 960
    Pack of 960 - $44.64
    $44.64
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 9600
    Case of 9600 - $446.40
    $446.40
    Prix promotionnel pour les nouveaux clients
    Pack of 960
       $44.64
    Case of 9600
       $446.40
    Supplier Number: 63300560
    Spécifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    89168-748
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
      Waste Reducing|Sustainable Packaging
  • Extended Length Tips
    1250 µl
    Clear
    96/Card
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 960
    Pack of 960 - $56.80
    $56.80
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 4800
    Case of 4800 - $284.00
    $284.00
    Prix promotionnel pour les nouveaux clients
    Pack of 960
       $56.80
    Case of 4800
       $284.00
    Supplier Number: 63300570
    Spécifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    89168-772
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
      Waste Reducing|Sustainable Packaging
  • Universal Fit Tips
    200 µl
    Clear
    96/Card
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 960
    Pack of 960 - $44.24
    $44.24
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 9600
    Case of 9600 - $442.40
    $442.40
    Prix promotionnel pour les nouveaux clients
    Pack of 960
       $44.24
    Case of 9600
       $442.40
    Supplier Number: 63300538
    Spécifications:
    Product available on GSA Advantage®
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    89140-902
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
    Product available on GSA Advantage®
      Waste Reducing|Sustainable Packaging
  • Universal Fit Tips
    300 µl
    Clear
    96/Card
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 960
    Pack of 960 - $44.64
    $44.64
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 9600
    Case of 9600 - $446.40
    $446.40
    Prix promotionnel pour les nouveaux clients
    Pack of 960
       $44.64
    Case of 9600
       $446.40
    Supplier Number: 63300523
    Spécifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    89168-742
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
      Waste Reducing|Sustainable Packaging
S3 Polymer Tips
Informations sur le produit
  • Tip Style
    Volume
    Tip Color
    For Use With
    Packed
    Availability
    Price
  • Extended Length Tips, Low Retention
    1250 µl
    Clear
    96/Card
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 960
    Pack of 960 - $60.88
    $60.88
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 4800
    Case of 4800 - $304.40
    $304.40
    Prix promotionnel pour les nouveaux clients
    Pack of 960
       $60.88
    Case of 4800
       $304.40
    Supplier Number: 63302179
    Spécifications:
    Product available on GSA Advantage®
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    89168-768
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
    Product available on GSA Advantage®
      Waste Reducing|Sustainable Packaging
  • Extended Length Tips, Universal Fit, Low Retention
    10 µl
    Clear
    96/Card
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 960
    Pack of 960 - $46.64
    $46.64
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 9600
    Case of 9600 - $466.40
    $466.40
    Prix promotionnel pour les nouveaux clients
    Pack of 960
       $46.64
    Case of 9600
       $466.40
    Supplier Number: 63302176
    Spécifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    89217-464
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
      Waste Reducing|Sustainable Packaging
  • Micro Tips, Low Retention
    10 µl
    Clear
    96/Card
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1,920
    Pack of 1,920 - $95.20
    $95.20
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 7680
    Case of 7680 - $380.80
    $380.80
    Prix promotionnel pour les nouveaux clients
    Pack of 1,920
       $95.20
    Case of 7680
       $380.80
    Supplier Number: 63302159
    Spécifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    89140-104
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
      Waste Reducing|Sustainable Packaging
  • Universal Fit Tips, Low Retention
    200 µl
    Clear
    96/Card
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 960
    Pack of 960 - $47.99
    $47.99
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 9600
    Case of 9600 - $479.88
    $479.88
    Prix promotionnel pour les nouveaux clients
    Pack of 960
       $47.99
    Case of 9600
       $479.88
    Supplier Number: 63302166
    Spécifications:
    Product available on GSA Advantage®
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    89140-900
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
    Product available on GSA Advantage®
      Waste Reducing|Sustainable Packaging
  • Universal Fit Tips, Low Retention
    300 µl
    Clear
    96/Card
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 960
    Pack of 960 - $45.44
    $45.44
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 9600
    Case of 9600 - $454.40
    $454.40
    Prix promotionnel pour les nouveaux clients
    Pack of 960
       $45.44
    Case of 9600
       $454.40
    Supplier Number: 63302162
    Spécifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    89168-738
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, ESP Reloads reduce plastic waste by 90%
      Waste Reducing|Sustainable Packaging
Neptune™ Empty Racks
Informations sur le produit
  • Tip Style
    Volume
    Tip Color
    For Use With
    Packed
    Availability
    Price
  • 10 - 300 µl tips, ESP reloads
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $23.76
    $23.76
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $118.80
    $118.80
    Prix promotionnel pour les nouveaux clients
    Pack of 10
       $23.76
    Case of 50
       $118.80
    Supplier Number: 63300801
    Spécifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    76311-242
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Empty rack intended for Reload and Bulk Refills
      Waste Reducing|Sustainable Packaging
  • 10 µl and 10 µl XL tips, bulk
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $28.90
    $28.90
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $144.51
    $144.51
    Prix promotionnel pour les nouveaux clients
    Pack of 10
       $28.90
    Case of 50
       $144.51
    Supplier Number: 63300800
    Spécifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    76311-290
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Empty rack intended for Reload and Bulk Refills
      Waste Reducing|Sustainable Packaging
  • 20 - 300 µl tips, bulk
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $28.73
    $28.73
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $143.67
    $143.67
    Prix promotionnel pour les nouveaux clients
    Pack of 10
       $28.73
    Case of 50
       $143.67
    Supplier Number: 63300802
    Spécifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    76311-244
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Empty rack intended for Reload and Bulk Refills
      Waste Reducing|Sustainable Packaging
  • 1000 - 1250 µl tips, ESP reloads
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 8
    Pack of 8 - $34.00
    $34.00
    Log in pour la disponibilité
    Case of 32
    Case of 32 - $136.00
    $136.00
    Pack of 8
       $34.00
    Case of 32
       $136.00
    Supplier Number: 63300803
    Spécifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    76311-246
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Empty rack intended for Reload and Bulk Refills
      Waste Reducing|Sustainable Packaging
  • 1000 - 1250 µl tips, bulk
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 8
    Pack of 8 - $39.90
    $39.90
    Log in pour la disponibilité
    Case of 32
    Case of 32 - $159.60
    $159.60
    Pack of 8
       $39.90
    Case of 32
       $159.60
    Supplier Number: 63300804
    Spécifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    76311-248
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Empty rack intended for Reload and Bulk Refills
      Waste Reducing|Sustainable Packaging