Axygen® Pipette Tip Reload System, Corning
Supplier: Corning
Certificates
About this item
Gilson®-style and universal fit pipette tips in easy-to-load refill racks.
- RNase-/DNase-free
- Nonpyrogenic
- Translucent rack allows tips to be easily seen
- Color-coded insert allows for easy identification of tip
Universal fit tips have a beveled reference and graduated marks.
Axygen® tip reload system contains durable refill rack that can be easily refilled by hand and is autoclavable. They are available in different colors and are packaged in bulk or racks.
Frequently Bought Together