About this item
Zebra Technologies® true colors card printer ribbons produce high-quality IDs and extend the life of your printers and cards. Vibrant colors, 'true-to-life' flesh tones, and sharp barcodes and text.
- For use with Zebra ZXP Series 3 card printers
- Ribbons are used for printing a combination of full-color images and one-color text or barcodes
- 200 color prints per roll
- Cleaning roller included
- OEM compatible
Patented print ribbon technology for minimal printhead buildup and durable cards. Zebra Technologies® True Colors I Series and C Series monochrome ribbons use thermal printer ribbon technology to deliver the most vibrant barcodes and text.
The ZXP 3 Load-N-Go cartridges makes it easy to load and replace the ribbon. They use intelligent media technology for automatic ribbon detection and optimized imaging. Optimize color card printer performance with a multi-panel thermal transfer ribbon offering the latest in dye-sublimation technology, resulting in crisp, long-lasting results.