Axygen® Research-Grade Pipette Tips, Corning
Supplier: Corning
Certificates
About this item
Tips are designed to fit a wide variety of single-channel and multichannel pipettors.
- RNase-, DNase-, and endotoxin-free
Presterilized tips are exposed to e-beam radiation for protection from contamination.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry.
Ordering information: Graduated tips have graduations at 10, 50, and 100 µl. The 200 µl wide-bore tips have a 1.52 mm (¹/₁₆") orifice, and 1000 µl wide-bore tips have a 2.0 mm ¹³/₁₆"
orifice. Both options for 30 µl tips and the 60 µl tips are designed for use with 16-channel pipettors for either 384- or 1536-well pipetting.
orifice. Both options for 30 µl tips and the 60 µl tips are designed for use with 16-channel pipettors for either 384- or 1536-well pipetting.
Specifications
- Catalog No:
- 10011-304
- 10011-312
- 10011-314
- 10011-316
- 10011-318
- 10011-928
- 10011-930
- 10011-932
- 10011-874
- 10011-876
- 10011-340
- 10011-344
- 10011-300
- 12000-742
- 47745-486
- 47745-484
- 10011-922
- 10011-326
- 10011-372
- 10011-374
- 10011-336
- 10011-338
- 12000-744
- 10011-320
- 10011-918
- 10011-322
- 10011-920
- 10011-324
- 47745-488
- 47734-724
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa