ContactUsLinkComponent
24-Well No-Bottom Plates, Greiner Bio-One
24-Well No-Bottom Plates, Greiner Bio-One
Catalog # 89085-338
Supplier:  Greiner Bio-One
Product available on GSA Advantage®
undefined
24-Well No-Bottom Plates, Greiner Bio-One
Catalog # 89085-338
Supplier:  Greiner Bio-One
Supplier Number:  662000-06
Product available on GSA Advantage®
Specifications
  • Color:
    Black
  • Cat. no.:
    89085-338
  • Supplier No.:
    662000-06
Specifications
About this item
Plates are made from virgin polystyrene resin and molded without the bottom of the plate.
  • Virgin polystyrene construction
  • Designed without a bottom
  • Chimney-well design prevents cross-contamination
  • Durable
They feature chimney-well design to help prevent cross-contamination.