Netwell™ Inserts, Sterile, Corning
Netwell™ Inserts, Sterile, Corning
   Options durables disponibles
Supplier:   Corning
Netwell™ Inserts, Sterile, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
Netwell™ Inserts, Sterile, Corning
4 Options Available Below
Product Details & Documents
Polyester mesh bottoms attached to PS rings or housing. Netwell™ inserts are used as tissue carriers, supports and strainers of small organs, tissue slices or explants at the air-media interface.
  • Handy carrier for immunocytochemical staining of tissue slices
  • Provides coarse filtration of tissue homogenates, cell suspensions and microcarriers
  • Available in two mesh sizes and diameters
  • Supplied sterile and pre-loaded in 6- or 12-well plates
  • 24 mm Netwell inserts fit in Corning® 50 ml plastic centrifuge tubes
Polystyrene reagent trays are available in black for improved visibility of tissue sections, or white for colorimetric reaction contrast. Specially designed carriers and handles permit the simultaneous processing of up to 12 samples per carrier. Each carrier kit contains eight carriers and eight handles.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 29442-132
  • 29442-136
  • 29442-138
  • 29442-134
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
Spécifications
Product Family Options
Informations sur le produit
  • Nb de puits
    Membrane Diameter
    Mesh Size
    Packaging
    Availability
    Price
  • 12
    15 mm
    74 µm
    12/Plate
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 48
    Case of 48 - $553.35
    $553.35
    Prix promotionnel pour les nouveaux clients
    Case of 48
       $553.35
    Supplier Number: 3477
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29442-132
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 6
    24 mm
    74 µm
    6/Plate
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 48
    Case of 48 - $651.10
    $651.10
    Prix promotionnel pour les nouveaux clients
    Case of 48
       $651.10
    Supplier Number: 3479
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29442-136
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 6
    24 mm
    440 µm
    6/Plate
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 48
    Case of 48 - $651.10
    $651.10
    Prix promotionnel pour les nouveaux clients
    Case of 48
       $651.10
    Supplier Number: 3480
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29442-138
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 12
    15 mm
    440 µm
    12/Plate
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 48
    Case of 48 - $592.80
    $592.80
    Prix promotionnel pour les nouveaux clients
    Case of 48
       $592.80
    Supplier Number: 3478
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29442-134
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
Accessories
Informations sur le produit
  • Catalog #
    Membrane Diameter
    Availability
    Price
  • Carrier kit, 6-well
    Carrier kit, 6-well
    29442-158
    24 mm
    Log in pour la disponibilité
    Case of 8 - $356.15
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Reagent Tray, Black
    Reagent Tray, Black
    29442-152
    Log in pour la disponibilité
    Case of 200 - $357.00
Informations sur le produit
  • Catalog #
    Membrane Diameter
    Availability
    Price
  • Carrier kit, 12-well
    Carrier kit, 12-well
    29442-156
    15 mm
    Log in pour la disponibilité
    Case of 8 - $398.05
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Reagent Tray, White
    Reagent Tray, White
    29442-154
    Log in pour la disponibilité
    Case of 200 - $420.00