epT.I.P.S.® Singles, Pipette Tips
   Options durables disponibles
Supplier:   Eppendorf
epT.I.P.S.® Singles, Pipette Tips
Supplier:  Eppendorf
   Options durables disponibles
Certificates
5 Options Available Below
Product Details & Documents
Eppendorf epT.I.P.S.® Singles high-precision pipette tips offer unparalleled precision in 'Biopur®' and 'Sterile' grades, featuring individually wrapped tips for maximum sterility. Ideal for sensitive and critical lab applications where contamination control is paramount.
  • Eppendorf pipette tip, individually wrapped
  • Available in Biopur and Sterile purity grades
  • For use in medical, pharmaceutical, food industry, molecular biology, and cell technology
  • Batch number and expiration date on each blister package
  • Each production lot inspected continuously by an independent laboratory
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally Integrated Pipetting System') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs.
Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.
How to recognize an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.
Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0.5 to 20 µl, 50 to 1250 µl, 0.2 to 5 ml, and 0.5 to 10 ml.
 : Lot-specific certficates for the purity grades Sterile, Biopur® can be downloaded from the Eppendorf website.
Specifications for Product Family
Spécifications
  • Catalog No:
  • CA47745-066
  • CA47745-068
  • CA47745-070
  • CA47745-218
  • CA47745-220
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Spécifications
Product Family Options
Biopur® (Sterile)
Informations sur le produit
  • Volume
    Sterile
    Longueur
    Color Code
    Packed
    Availability
    Price
  • 0.1 - 20 µl
    +
    40 mm
    Light orange
    100 Tips, individually Packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $74.70
    $74.70
    Case of 100
       $74.70
    Supplier Number: 022491130
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA47745-066
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 2 - 200 µl
    +
    53 mm
    Yellow
    100 Tips, individually Packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $64.60
    $64.60
    Case of 100
       $64.60
    Supplier Number: 022491148
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA47745-068
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 50 - 1000 µl
    +
    71 mm
    Blue
    100 Tips, individually Packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $65.80
    $65.80
    Case of 100
       $65.80
    Supplier Number: 022491156
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA47745-070
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
Sterile
Informations sur le produit
  • Volume
    Sterile
    Longueur
    Color Code
    Packed
    Availability
    Price
  • 2 - 200 µl
    +
    53 mm
    Yellow
    1,000 Tips, individually Packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $340.00
    $340.00
    Case of 1,000
       $340.00
    Supplier Number: 022492209
    Spécifications:
    More
    Specifications
    Cat. No.
    CA47745-218
    Environmentally Preferable
    Environmental certification
  • 50 - 1000 µl
    +
    71 mm
    Blue
    1,000 Tips, individually Packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $340.00
    $340.00
    Case of 1,000
       $340.00
    Supplier Number: 022492225
    Spécifications:
    More
    Specifications
    Cat. No.
    CA47745-220
    Environmentally Preferable
    Environmental certification