Nalgene® Safety Bottle Carriers, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
For the transport and storage of stockroom chemicals, glass bottles, and field samples
Clear plastic lid permits identification of color-coded closures on acid bottles and protects the neck and bottle cap from damage. Lid may be removed with a quick twist; pour directly from the bottle without removal from the carrier.
Carriers are manufactured of impact-resistant, low-density polyethylene and hold metric-sized chemical bottles for improved safety and ease of handling. Bottles with fingerloops fit easily.
Specifications
- Catalog No:
- 56609-200
- 56609-222
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)