96-Well Microplates, Polystyrene, Clear, Greiner Bio-One
96-Well Microplates, Polystyrene, Clear, Greiner Bio-One
96-Well Microplates, Polystyrene, Clear, Greiner Bio-One
Supplier:  Greiner Bio-One
Certificates
96-Well Microplates, Polystyrene, Clear, Greiner Bio-One
96-Well Microplates, Polystyrene, Clear, Greiner Bio-One
13 Options Available Below
Product Details & Documents
These 96-well microplates are available with flat-, round-, conical-, and C-bottom well styles, and are manufactured from clear, virgin polystyrene and selected for optical clarity and consistency.
  • Clear virgin polystyrene construction
  • High degree of optical clarity
  • Chimney-well design helps reduce cross-contamination
  • Designed to minimize air bubble formation
C-bottom plates feature a flat bottom for optical measurement, but the hard corners are curved to eliminate liquid traps and improve plate washing
All plates are free of detectable RNase, DNase, human DNA, and pyrogens.
MICROLON™ 200 and MICROLON™ 600 formats are available for consistent performance in binding assays. The chimney-well design option reduces the risk of cross-contamination and facilitates sealing by cap mat, thermal sealer, or adhesive sealer. Plates are supplied without lids.
Documents
Product Family Options
MICROLON™ 200 Medium Binding 96-Well Microplates
Informations sur le produit
  • Color
    Surface Treatment
    Well Shape
    Availability
    Price
  • Clear
    Medium binding
    Conical-Bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 40
    Case of 40 - $205.00
    $205.00
    Case of 40
       $205.00
    Supplier Number: 651001
  • Clear
    Medium binding
    Flat-Bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 40
    Case of 40 - $172.00
    $172.00
    Case of 40
       $172.00
    Supplier Number: 655001
  • Clear
    Medium binding
    Flat-Bottom, with Chimney
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 40
    Case of 40 - $172.00
    $172.00
    Case of 40
       $172.00
    Supplier Number: 655080
  • Clear
    Medium binding
    Round-Bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 40
    Case of 40 - $150.00
    $150.00
    Case of 40
       $150.00
    Supplier Number: 650001
MICROLON™ 600 High Binding 96-Well Microplates
Informations sur le produit
  • Color
    Surface Treatment
    Well Shape
    Availability
    Price
  • Clear
    High binding
    Conical-Bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 40
    Case of 40 - $207.00
    $207.00
    Case of 40
       $207.00
    Supplier Number: 651061
  • Clear
    High binding
    C-Bottom, with Chimney
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 40
    Case of 40 - $187.18
    $187.18
    Prix promotionnel pour les nouveaux clients
    Case of 40
       $187.18
    Supplier Number: 655085
  • Clear
    High binding
    Flat-Bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 40
    Case of 40 - $167.98
    $167.98
    Prix promotionnel pour les nouveaux clients
    Case of 40
       $167.98
    Supplier Number: 655061
  • Clear
    High binding
    Flat-Bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 150
    Case of 150 - $674.60
    $674.60
    Prix promotionnel pour les nouveaux clients
    Case of 150
       $674.60
    Supplier Number: 655061-25
  • Clear
    High binding
    Flat-Bottom, with Chimney
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 40
    Case of 40 - $168.00
    $168.00
    Case of 40
       $168.00
    Supplier Number: 655081
  • Clear
    High binding
    Round-Bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 40
    Case of 40 - $158.00
    $158.00
    Case of 40
       $158.00
    Supplier Number: 650061
Standard 96-Well Microplates
Informations sur le produit
  • Color
    Surface Treatment
    Well Shape
    Availability
    Price
  • Clear
    Untreated
    Conical-Bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $181.00
    $181.00
    Case of 100
       $181.00
    Supplier Number: 651101
  • Clear
    Untreated
    Flat-Bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $186.00
    $186.00
    Case of 100
       $186.00
    Supplier Number: 655101
  • Clear
    Untreated
    Round-Bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $191.00
    $191.00
    Case of 100
       $191.00
    Supplier Number: 650101