Storage Microplates 384-well Polypropylene, Corning®
À propos de cet article
These 384-well polypropylene plates and blocks are built to the industry standard footprint
Robolid combines the sealing ability of a storage mat with the rigidity of a plastic lid and is designed for repeated application and removal by automation and to prevent short-term evaporation. Robolid also features silicone sealing plugs for organic solvent resistance and low extractables.
They are automation-friendly and solvent-resistant. They feature round-, conical-, or v-bottoms for optimal liquid handling and a rounded square-well geometry for reduced wicking. Both plates and blocks are certified DNase- and RNase-free.
Ordering information: The 384-well plates are packaged 25 per pack, except for 46610-064 which is packaged 10 per pack. The 384-well blocks are packaged 5 per pack.
7 Options Available Below
Product Details & Documents
These 384-well polypropylene plates and blocks are built to the industry standard footprint
Robolid combines the sealing ability of a storage mat with the rigidity of a plastic lid and is designed for repeated application and removal by automation and to prevent short-term evaporation. Robolid also features silicone sealing plugs for organic solvent resistance and low extractables.
They are automation-friendly and solvent-resistant. They feature round-, conical-, or v-bottoms for optimal liquid handling and a rounded square-well geometry for reduced wicking. Both plates and blocks are certified DNase- and RNase-free.
Ordering information: The 384-well plates are packaged 25 per pack, except for 46610-064 which is packaged 10 per pack. The 384-well blocks are packaged 5 per pack.
Specifications for Product Family
Spécifications
- Catalog No:
- 15551-254
- 29445-120
- 29445-136
- 29445-114
- 29445-116
- 80081-116
- 80081-126
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Spécifications
Product Family Options
384-Well Plates
Informations sur le produit
- DésignationWell VolumeColorSterilityAvailabilityPrice
- Round-Bottom95 µLBlackNon sterileLog in pour la disponibilitéLog in pour la disponibilitéCase of 100Case of 100 - $986.00$986.00Prix promotionnel pour les nouveaux clientsCase of 100$986.00Supplier Number: 3658Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.15551-254Environmentally PreferableEnvironmental certificationProduct available on GSA Advantage® - Round-Bottom95 µLClearSterileLog in pour la disponibilitéLog in pour la disponibilitéCase of 100Case of 100 - $752.25$752.25Prix promotionnel pour les nouveaux clientsCase of 100$752.25Supplier Number: 3656Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.29445-120Environmentally PreferableEnvironmental certificationProduct available on GSA Advantage® - Round-Bottom95 µLClearNon sterileLog in pour la disponibilitéLog in pour la disponibilitéCase of 100Case of 100 - $674.05$674.05Prix promotionnel pour les nouveaux clientsCase of 100$674.05Supplier Number: 3657Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.29445-136Environmentally PreferableEnvironmental certificationProduct available on GSA Advantage®
384-Well Blocks
Informations sur le produit
- DésignationWell VolumeColorSterilityAvailabilityPrice
- Supplier Number: 3965Spécifications:
More
Specifications
Cat. No.29445-114Environmentally PreferableEnvironmental certification - Round-Bottom180 µLClearSterileLog in pour la disponibilitéLog in pour la disponibilitéCase of 25Case of 25 - $388.45$388.45Prix promotionnel pour les nouveaux clientsCase of 25$388.45Supplier Number: 3964Spécifications:Product available on GSA Advantage®
More
Specifications
Cat. No.29445-116Environmentally PreferableEnvironmental certificationProduct available on GSA Advantage® - V-Bottom240 µLClearSterileLog in pour la disponibilitéLog in pour la disponibilitéCase of 50Case of 50 - $613.70$613.70Prix promotionnel pour les nouveaux clientsCase of 50$613.70Supplier Number: 3342Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.80081-116Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Reynosa - V-Bottom240 µLClearNon sterileLog in pour la disponibilitéLog in pour la disponibilitéCase of 50Case of 50 - $510.00$510.00Prix promotionnel pour les nouveaux clientsCase of 50$510.00Supplier Number: 3347Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.80081-126Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Reynosa
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Sealing Mat