Microlite™ Luminescence Microtiter® 96-Well Plates, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
These plates were designed for applications in luminescence.
Microlite™ 1 is ideal for applications requiring high reflectivity and minimum crosstalk. Microlite™ 2, irradiated, is ideal for applications requiring higher binding with the same high reflectivity and minimum crosstalk. Plates are compatible with Thermo Scientific universal microplate lids (see 62402-929 series) which are available individually wrapped or sterile.
These plates are made of polystyrene.
Frequently Bought Together