Nunc® 96-Well Filter Plates, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
Filter plates are made of polypropylene, which offers chemical resistance and mechanical strength
- Untreated
They are designed specifically for high throughput sample processing, and are also suitable for vacuum filtration, centrifugation, or positive pressure techniques. The filter plates have a shred wall configuration which gives a reduced plate height, wider wells for faster processing, higher yields, and less clogging
Certifications: Microplates meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004).
Frequently Bought Together