VWR® Traceable® Bench/Portable Conductivity Meter
VWR® Traceable® Bench/Portable Conductivity Meter
Benchtop/portable conductivity meter
Catalog # 23226-505
Supplier:   VWR International
VWR® Traceable® Bench/Portable Conductivity Meter
Catalog # 23226-505
Supplier:  VWR International
VWR® Traceable® Bench/Portable Conductivity Meter
VWR® Traceable® Bench/Portable Conductivity Meter
Benchtop/portable conductivity meter
Price
$4,459.05
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Designed to measure conductivity in water analysis, biology, chromatography, food, electronics, dairies, and PC-board rinsing, the conductivity meter provides results in conductivity (micromhos/cm and microsiemens/cm), resistivity (megohms), and dissolved solids (parts per million).
  • Traceable®
The tough, chemical-resistant ABS housing withstands the harshest environments. The 12.7 mm (¹/₂") high 4-digit LCD readout shows temperature in °C or °F. Recorder jack allows continuous monitoring and a permanent record. The epoxy conductivity probe has a range of 1.0–200000 Micromhos.
Certifications: Meets all accreditation requirements and U.S. Government specifications. Measures reagent grade water for CAP, ASTM, NCCLS, and ACS. A serial numbered certificate is provided from an ISO 17025 calibration laboratory to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Unit complete with glass probe and 9 V alkaline battery.
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Spécifications
  • Désignation:
    Benchtop/portable conductivity meter
  • Cat. No.:
    23226-505
  • Conductivity accuracy:
    0.3%
  • Conductivity range:
    0.01–20 µS, 0.1–200 µS, 1–2000 µS, 10–20,000 µS, 100–200,000 µS, 0.001–2 megohms, 0.01–20 megohms, 0.1–20 ppm dissolved solids, 1–200 ppm dissolved solids, 10–2000 ppm dissolved solids, 100–20,000 ppm dissolved solids
  • Dimensions:
    9.5L x 3.8D x 17.1H cm (33/4 x 11/2 x 63/4")
  • Temperature range:
    –30.0 to 130.0°C (–22 to 266°F)
  • Probe:
    1.27 dia. x 13.3L cm (1/2 x 51/4")
  • Weight:
    453.6 g (16 oz.)
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement glass probe
    Replacement glass probe
    23226-504
    Log in pour la disponibilité
    Each - $925.35