Eppendorf® Xplorer® plus multi-channel electronic pipettes offer the ideal solution for precise, efficient liquid handling in high-throughput applications, with ergonomic design, a user-friendly interface, and programmable modes for quick, reliable pipetting, enhancing reproducibility and minimizing errors.
- Multifunctional rocker: 'Up is up and down is down™' principle
- Clearly arranged color display; no sub-menus; help function
- Innovative rocker: When the tip is ejected, the piston automatically returns to zero position
- ON/OFF switch: Switches the pipetter off during longer periods of inactivity
- Programming: One-time programming of sequences for easier handling of complex processes
- Individual fixed volume setting: Desired volume can be saved
- Password protection: Protects programming and adjustment
- Individual speed setting: Sets parameters to your individual working speed
Reliably reproducible results:
Tips always rest in the same position
Volume and speed precisely and reproducibly adjustable
Adjustable to your work environment and requirements
PhysioCare Concept™:
Reduced weight
Improved ergonomics
Perfect balance
Optimally angled pipetter head
Improved view and high resolution display
Spring-loaded tip cone provides maximum tightness
Pipetter maintenance reminder function, based on time or frequency of use as desired. Operate in the language of your choice. And has an easy selection of all functions with the practical selection dial.
Air-cushion pipetting system, with autoclavable lower part.
Applications: Pipetting of aqueous liquids, pipetting liquids (manual and automatic), dispensing (timed and manual) and pipetting and mixing. Additional applications include aspirating, diluting, sequential dispensing and reverse pipetting.
- Catalog No:
- CA89234-008
- CA89234-010
- CA89234-016
- CA89234-018
- CA89234-012
- CA89234-014
- CA76311-136
- CA89234-020
- CA76311-130
- CA89234-022
- CA76311-132
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001