Spunbond Coveralls
Fournisseur: O&M HALYARD CANADA, INC CA
New Product
Certificats
À propos de cet article
HALYARD* Spunbond coveralls are manufactured with lightweight polypropylene spunbond fabric for breathability and wearer's comfort. These coveralls are appropriate for situations in which light fluid contact or contact isolation is anticipated.
- For low exposure to fluids or chemicals
- Spunbond fabric
- Knit cuffs
- Open legs
- Full zip front
Spécifications
- Catalog No:
- 77531-674
- 77531-672
- 77531-678
- 77531-676
- Fabric:
- Spunbond Polypropylene
- Spunbond Polypropylene
- Spunbond Polypropylene
- Spunbond Polypropylene
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Seam type:
- Stitched
- Stitched
- Stitched
- Stitched
- Garment application:
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection