Protective Coveralls | Avantor
Protective Coveralls
Protective Coveralls
Certificates
3 Options Available Below
Product Details & Documents
HALYARD* Protective coveralls are appropriate for situations in which light fluid contact or contact isolation is anticipated. Features include a soft breathable 3-layer SMS fabric plus tailored styling and fit to enhance worker comfort.
  • Raglan sleeve
  • Protective 3-layer SMS fabric
  • For low exposure to fluids or chemicals
  • Traditional collar
  • Single use
  • Recommended areas for product use: Central services construction cleaning and maintenance
Specifications for Product Family
Spécifications
  • Catalog No:
  • 77531-666
  • 77531-668
  • 77531-670
  • Fabric:
  • 3-layer SMS fabric
  • 3-layer SMS fabric
  • 3-layer SMS fabric
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Pockets:
  • Without pockets
  • Without pockets
  • Without pockets
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Seam type:
  • Stitched
  • Stitched
  • Stitched
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
Spécifications
Product Family Options
Informations sur le produit
  • Color
    Taille de confection
    Longueur
    Availability
    Price
  • Blue
    L
    170.2 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $252.37
    $252.37
    Case of 24
       $252.37
    Supplier Number: 75631
    Spécifications:
    More
    Specifications
    Cat. No.
    77531-666
    Fabric
    3-layer SMS fabric
    Closure type
    Zip closure
    Pockets
    Without pockets
    Sleeve or leg length
    Long
    Seam type
    Stitched
    Garment application
    Cleanroom
  • Blue
    X-Large
    170.2 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $282.07
    $282.07
    Case of 24
       $282.07
    Supplier Number: 75641
    Spécifications:
    More
    Specifications
    Cat. No.
    77531-668
    Fabric
    3-layer SMS fabric
    Closure type
    Zip closure
    Pockets
    Without pockets
    Sleeve or leg length
    Long
    Seam type
    Stitched
    Garment application
    Cleanroom
  • Blue
    XX-Large
    170.2 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $282.03
    $282.03
    Case of 24
       $282.03
    Supplier Number: 75651
    Spécifications:
    More
    Specifications
    Cat. No.
    77531-670
    Fabric
    3-layer SMS fabric
    Closure type
    Zip closure
    Pockets
    Without pockets
    Sleeve or leg length
    Long
    Seam type
    Stitched
    Garment application
    Cleanroom