Wheatley Trichrome solution réactif de coloration
Wheatley Trichrome solution réactif de coloration
Plastic bottle, 250 ml
Catalog # 77540-316
Supplier:   G-Biosciences
Supplier #:  786-2129
Wheatley Trichrome solution réactif de coloration
Catalog # 77540-316
Supplier:  G-Biosciences
CAS Number:  
Wheatley Trichrome solution réactif de coloration
Plastic bottle, 250 ml
CAS Number:  
Price
$155.76
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Wheatley Trichrome quick stain is recommended for detection and differentiation of intestinal protozoa.
  • Quick staining dye
Spécifications
Spécifications
  • Type de conditionnement:
    Plastic bottle
  • Taille:
    250 ml