Wheatley Trichrome kit de coloration
Plastic bottle, 1 kit
Catalog # 77540-314
Supplier:   G-Biosciences
 :  786-2128
Wheatley Trichrome kit de coloration
Catalog # 77540-314
Supplier:  G-Biosciences
CAS Number:  
Wheatley Trichrome kit de coloration
Plastic bottle, 1 kit
CAS Number:  
Price
$777.22
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Wheatley Trichrome quick stain kit is a complete set of reagents recommended for detection and differentiation of intestinal protozoa.
  • Staining kit
 : Kit includes Wheatley Trichrome Stain (1 x 250 ml), Xylene S (2 x 250 ml), Ethanol 90% (2 x 250 ml) and Acid Ethanol 90% (1 x 250 ml), Ethanol 70% (4 x 250 ml) and Lugol’s Iodine (5 x 1.0 ml)
Spécifications
Spécifications
  • Type de conditionnement:
    Plastic bottle
  • Taille:
    1 kit