Wind Tunnel
Wind Tunnel
Wind tunnel for educational demonstration
Catalog # 470356-464
Supplier:   GSC International
Supplier #:  WNDTNL
Wind Tunnel
Catalog # 470356-464
Supplier:  GSC International
Wind Tunnel
Wind Tunnel
Wind tunnel for educational demonstration
Price
102,70 $
Log in pour la disponibilité
Each
Product Details & Documents
Ideal for investigating lift, Bernouli's Principle, and aerodynamics.
Simply connect the tunnel to a power source (not included), turn the fan on and place a source of smoke in front of the fan. You will be able to watch the smoke move at different rates on either side of the wing.
Ordering information: Power source and source of smoke are not included.
Caution: Use in a laboratory or educational setting only. Not for children 13 and under.
Spécifications
  • Désignation:
    Wind tunnel for educational demonstration