ContactUsLinkComponent
Acrodisc™ Syringe Filters with Nylon Membrane, Cytiva (Formerly Pall Lab)
Acrodisc™ Syringe Filters with Nylon Membrane, Cytiva (Formerly Pall Lab)
Supplier:  Cytiva
New Product
Certificates
undefined
Acrodisc™ Syringe Filters with Nylon Membrane, Cytiva (Formerly Pall Lab)
Supplier:  Cytiva
New Product
Specifications
  • Catalog No:
  • CA77553-278
  • CA77553-276
  • CA77553-298
  • CA77553-308
  • Type:
  • CA77553-278
  • CA77553-276
  • CA77553-298
  • CA77553-308
  • Hold-up volume:
  • <125 µl
  • <425 µl
  • <14 µl
  • <125 µl
  • Max. operating pressure:
  • 30.4 psi
  • 30.4 psi
  • 100 psi
  • 30.4 psi
  • Max. temperature:
  • 55 °C
  • 55 °C
  • 55 °C
  • 55 °C
  • Water flow rate:
  • 115 ml/min at 2.1 bar (210 kPa, 30 psi)
  • 215 ml/min at 2.1 bar (210 kPa, 30 psi)
  • 15 ml/min at 2.1 bar (210 kPa, 30 psi)
  • 15 ml/min at 2.1 bar (210 kPa, 30 psi)