À propos de cet article
Acrodisc™ syringe filters with PTFE membrane provide exceptional chemical and temperature compatibility.
- Ideal for filtration of gas and/or organic solvents
- Prevents spurious peaks on chromatograms for accurate experimental results
- HPLC certified for low levels of UV-absorbing extractables
- Prevents 'air bubble' lock when filling sample vials with 13 mm minispike outlet
The device offers the lowest sample hold-up of less than 10 μl and is useful for samples up to 2 ml in volume.
The Acrodisc™ PSF GxF syringe filter with PTFE membrane offers ultimate chemical compatibility, making it ideal for filtering harsh chemicals that can destroy other membrane materials. It provides two to four times the throughput of standard prefilter devices, and serves as an effective moisture barrier for venting applications. Additionally, the 25 mm Acrodisc™ PSF syringe filter is available in robotic-compatible AutoPack™ packaging, enhancing its suitability for automated processes.
The Acrodisc syringe filters feature a hydrophobic PTFE membrane on a polypropylene support.
Spécifications
- Catalog No:
- CA77553-288
- CA77553-314
- CA77553-286
- CA77553-284
- CA77553-296
- CA77553-312
- Type:
- CA77553-288
- CA77553-314
- CA77553-286
- CA77553-284
- CA77553-296
- CA77553-312
- Hold-up volume:
- <125 µl
- <125 µl
- <125 µl
- <125 µl
- <14 µl
- <125 µl
- Max. operating pressure:
- 30.4 psi
- 30.4 psi
- 59.4 psi
- 30.4 psi
- 100 psi
- 30.4 psi
- Max. temperature:
- 100 °C
- 100 °C
- 55 °C
- 100 °C
- 100 °C
- 100 °C
- Water flow rate:
- 510 ml/min at 2.1 bar (210 kPa, 30 psi)
- 510 ml/min at 1.0 bar (100 kPa, 15 psi)
- 215 ml/min at 2.1 bar (210 kPa, 30 psi)
- 395 ml/min at 1.0 bar (100 kPa, 15 psi)
- 510 ml/min at 1.0 bar (100 kPa, 15 psi)
- 245 ml/min at 1.0 bar (100 kPa, 15 psi)