ContactUsLinkComponent
Acrodisc™ Syringe Filters with Universal Membranes, Cytiva (Formerly Pall Lab)
Acrodisc™ Syringe Filters with Universal Membranes, Cytiva (Formerly Pall Lab)
Supplier:  Cytiva
New Product
Certificates
undefined
Acrodisc™ Syringe Filters with Universal Membranes, Cytiva (Formerly Pall Lab)
Supplier:  Cytiva
New Product
Specifications
  • Catalog No:
  • CA77553-318
  • CA77553-316
  • CA77553-294
  • CA77553-270
  • CA77553-268
  • CA77553-272
  • Hold-up volume:
  • <14 µl
  • <14 µl
  • <100 µl
  • <125 µl
  • <125 µl
  • <125 µl
  • Max. operating pressure:
  • 89.9 psi
  • 89.9 psi
  • 89.9 psi
  • 89.9 psi
  • 89.9 psi
  • 89.9 psi
  • Max. temperature:
  • 55 °C
  • 55 °C
  • 55 °C
  • 55 °C
  • 55 °C
  • 55 °C
  • Water flow rate:
  • 27 ml/min at 1.0 bar (100 kPa, 15 psi)
  • 51 ml/min at 1.0 bar (100 kPa, 15 psi)
  • 262 ml/min at 1.0 bar (100 kPa, 15 psi)
  • 263 ml/min at 1.0 bar (100 kPa, 15 psi)
  • 224 ml/min at 1.0 bar (100 kPa, 15 psi)
  • 151 ml/min at 1.0 bar (100 kPa, 15 psi)