Corning® Primaria™ Culture Plates, Corning
Supplier:   Corning
Corning® Primaria™ Culture Plates, Corning
Supplier:  Corning
Certificates
Corning® Primaria™ Culture Plates, Corning
Corning® Primaria™ Culture Plates, Corning
3 Options Available Below
Product Details & Documents
Unique nitrogen-containing tissue culture surface chemistry. Improves attachment, spreading, and growth for many primary cells or cell lines. Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process.
  • Stable, permanent surface modification
  • Optically clear
  • No special storage required
  • Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
  • Packaged in red color-coded, peel-open, medical-style packages
  • Sterilized by gamma irradiation
  • Nonpyrogenic tested to less than 0.1 EU/ml
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Sterility
    Packaging
    Availability
    Price
  • 6-Well
    PS
    Sterile
    1/Tray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $373.00
    $373.00
    Case of 50
       $373.00
    Supplier Number: 353846
  • 24-Well
    PS
    Sterile
    1/Tray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $477.00
    $477.00
    Case of 50
       $477.00
    Supplier Number: 353847
  • 96-Well
    PS
    Sterile
    1/Tray
    Discontinued
    Supplier Number: -
  • 96-Well
    PS
    Sterile
    1/Tray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $513.40
    $513.40
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Each
    Each - $513.40
    $513.40
    Prix promotionnel pour les nouveaux clients
    Case of 50
       $513.40
    Each
       $513.40
    Supplier Number: 353872