ContactUsLinkComponent
ART® 5000 Self-Sealing Barrier Pipette Tips, Molecular BioProducts
ART® 5000 Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Catalog # 17810-498
Supplier:  Thermo Scientific
   Low Manufacturing Impact
undefined
ART® 5000 Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Catalog # 17810-498
Supplier:  Thermo Scientific
Supplier Number:  2180B
   Low Manufacturing Impact
Specifications
  • Volume:
    1000 - 5000 µL
  • Length:
    11.7 cm (4⅝")
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO14001 (Certificate Registration No. EMS 752603)
  • Cat. no.:
    17810-498
  • Supplier No.:
    2180B
Specifications
About this item
Designed with a patented, self-sealing barrier, ART tips are the only tips that completely block the passage of aqueous based liquids and aerosols from the sample to the pipettor.
  • All tips are pre-sterilized
ART tips are designed to draw and dispense the full volume of the tip and the fine point design is an additional feature to ensure that samples can be drawn and dispensed with accuracy.
Tips are compatible with Pipetman® P-5000, and Nichiryo/Benchmate® series pipettors.
The unique seal protects the pipetting device from any possible contamination that could carry over and compromise future samples. ART tips are ideal for critical uses such as PCR, sequencing, loading gels, handling infectious samples and pipetting radioactive isotopes. The barrier formed by an ART filter is so secure it even prevents 7.2 million counts of 32P from breaking through. The purity of ART tips is achieved by constant control against contamination from any source. Electron beam sterilization is used because it ensures total disruption of any molecules amplifiable by PCR.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Packaging: Sterile tips are bulk packed.