ART® 1000E Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Fournisseur: Thermo Scientific
Certificats
À propos de cet article
Tips ensure 100% protection against aerosol and liquid contamination.
- Pre-sterilized tips
- Tips provide an ideal fit for most 1000 µl pipettors
The ART® barrier traps contaminants, keeping them out of pipettors and samples. This also protects the sensitive inner components of the pipettor, prolonging pipettor life. ART® tips are ideal for nucleic amplification procedures, pipetting of radioactive samples, or any application where critical sample handling is required.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Packaging: Sterile tips are packaged 100/tray, 8 trays per pack. Non sterile tips are bulk packed. The case of 500 tips are individually wrapped.
Spécifications
- Catalog No:
- 53225-782
- 72830-042
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)