ART® XLP Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Catalog # 72830-026
Supplier: Thermo Scientific
Specifications
- Volume:180 µL
- Length:9.12 cm (3¹⁹⁄₃₂")
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Cat. no.:72830-026
- Sterile:+
- Supplier No.:2159P
Specifications
About this item
Tips ensure 100% protection against aerosol and liquid contamination.
- Pre-sterilized tips
The extended length of these tips makes it easier to reach the bottom of 12×75 culture tubes and vials. The ART® barrier traps contaminants, keeping them out of pipettors and samples. This also protects the sensitive inner components of the pipettor, prolonging pipettor life. ART® tips are ideal for nucleic amplification procedures, pipetting of radioactive samples, or any application where critical sample handling is required. Tips are compatible with Eppendorf®, Finnpipette®, Pipetman®, and Rainin® series pipettors.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Packaging: Sterile tips are packaged 96/tray, 8 trays per pack. Non sterile tips are bulk packed.