Nunc® Rectangular Dishes, Polystyrene, Sterile, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
These 1-, 4-, and 8-well rectangular dishes feature an untreated surface for molecular biology applications
The 1-well dish is ideal for HTS petri dish applications. The 4- and 8-well dishes are ideal for compartmentalizing assays, probing multiple gene subsets, and washing Western blot strips. The 4-well dish can hold four standard glass slides. All dishes are made of polystyrene and feature ANSI standard dimensions for instrument compatibility. Sterile. Color: clear.
Ordering information: All dishes include lids.
Frequently Bought Together