Trespa® Toplab® Countertops, AGR Fabricators
Supplier: AGR
Certificates
About this item
These compression-molded countertops are composed of thermoset resins with cellulose fiber reinforcement for outstanding resistance to chemicals, heat, and stains.
- The nonporous tops will not rot or rust and are resistant to fungus, bacteria, and UV rays
- Durable tops can be routinely disinfected and do not require surface dressing
- Ease of machinability permits subsequent field modifications for equipment, piping, and wiring
Curved and other non-standard shaped tops are also possible. Countertops have a black core and black surface. Thickness: 2.5 cm (1").
Ordering information: Surface colors including light gray, light beige, and white are also available; contact your Avantor sales representative for more information.
Frequently Bought Together