Pipette Tips for Rainin LiteTouch™ Pipettors, Molecular BioProducts
ART® self-sealing barriers trap contaminants, such as aerosols and liquids, keeping them out of pipettors and samples.
- ART® tips are presterilized
- Several tips are available with low retention technology
Tips are available in non sterile packaging, in sterile packaging, or with ART self-sealing barrier.
This allows the user to conduct PCR procedures and pipette radioactive or infectious samples. This permits more accurate sample dispensing, an increased level of reproducibility, and reduced sample loss due to increased adhesion to pipette tips.
: Certified RNase-, DNase-, DNA-, and pyrogen-free.
: All tips are packaged 96/rack.
- Catalog No:
- 72830-444
- 72830-284
- 72830-471
- 82031-392
- 72830-454
- 72830-270
- 72830-268
- 72830-278
- 72830-276
- 72830-272
- 72830-282
- 72830-280
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
Frequently Bought Together