Nunclon™ Delta Rectangular Dishes, Polystyrene, Sterile, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
These 4- and 8-well rectangular dishes feature the certified Nunclon™Δ surface for consistent cell attachment and growth
The dishes are useful in all areas of cell culture, including scale-up and cloning. They are ideal for compartmentalizing cell culture assays. The 4-well dish can hold four standard glass slides. Both dishes are made of polystyrene and feature ANSI standard dimensions for instrument compatibility. Sterile. Color: clear.
Ordering information: Both dishes include lids.
Frequently Bought Together