LE510 Redox Combination Electrode, METTLER TOLEDO®
CA97002-312
- Temperature Range:0...80 °C
- Length:120 mm
- Material:Plastic
- Cat. no.:CA97002-312
- Junction:Ceramic
- mV Range:±2000 mV
Gel-filled electrode is for use with METTLER TOLEDO® pH meters in water analysis.
- Gell-filled
- Plastic body
- BNC connector
Robust plastic body makes it ideal for field measurements. Features BNC connector.