polyproLABS® Wall Cabinets, Air Control
Corrosion-resistant casework cabinets are constructed of stress-relieved, acid-resistant, 1.3 cm (1/2") thick polypropylene
Frameless acrylic sliding doors are 1 cm (3/8") thick. Framed acrylic doors have polypropylene frames and 1 cm (3/8") thick acrylic panels with beveled edges. Solid polypropylene doors are 1 cm (3/8") thick with beveled edges.
Cabinets include an adjustable shelf, which is made from solid 1.3 cm (1/2") thick polypropylene. Blind corner cabinets do not include a shelf. Open-front bookcase cabinets include two adjustable shelves.
Joints are thermally welded by a special process that ensures leak-tight, flush unions. All hardware, including door hinges, screws, latches, and handles, is constructed of polypropylene. No metals are used in cabinets.
Corrosion-resistant casework cabinets are constructed of stress-relieved, acid-resistant, 1.3 cm (1/2") thick polypropylene
Frameless acrylic sliding doors are 1 cm (3/8") thick. Framed acrylic doors have polypropylene frames and 1 cm (3/8") thick acrylic panels with beveled edges. Solid polypropylene doors are 1 cm (3/8") thick with beveled edges.
Cabinets include an adjustable shelf, which is made from solid 1.3 cm (1/2") thick polypropylene. Blind corner cabinets do not include a shelf. Open-front bookcase cabinets include two adjustable shelves.
Joints are thermally welded by a special process that ensures leak-tight, flush unions. All hardware, including door hinges, screws, latches, and handles, is constructed of polypropylene. No metals are used in cabinets.
Product Family Options
- DésignationMatièreLargeurProfondeurHauteurWeight
- Base cabinets, 2 frameless acrylic sliding doorsPP36"13"30"100 lbs.Each$4,545.80: AC-240-3030
Specifications
Cat. No.89008-514 - Base cabinets, 2 frameless acrylic sliding doorsPP42"13"30"125 lbs.Each$4,669.07: AC-240-3630
Specifications
Cat. No.89008-516 - Base cabinets, 2 frameless acrylic sliding doorsPP48"13"30"150 lbs.Each$4,796.75: AC-240-4230
Specifications
Cat. No.89008-518 - Base cabinets, 2 frameless acrylic sliding doorsPP30"13"24"175 lbs.Each$4,924.62: AC-240-4830
Specifications
Cat. No.89008-520 - Base cabinets, 2 frameless acrylic sliding doorsPP36"13"24"80 lbs.Each$4,669.07: AC-240-3024
Specifications
Cat. No.89008-522 - Base cabinets, 2 frameless acrylic sliding doorsPP42"13"24"100 lbs.Each$4,796.75: AC-240-3624
Specifications
Cat. No.89008-524 - Base cabinets, 2 frameless acrylic sliding doorsPP48"13"24"120 lbs.Each$4,924.62: AC-240-4224
Specifications
Cat. No.89008-526 - Base cabinets, 2 frameless acrylic sliding doorsPP30"13"30"140 lbs.Each$4,924.62: AC-240-4824
Specifications
Cat. No.89008-528 - Base cabinets, 2 framed acrylic sliding doorsPP30"13"48"110 lbs.Each$4,545.80: AC-250-3030
Specifications
Cat. No.89008-530 - Base cabinets, 2 framed acrylic sliding doorsPP36"13"30"185 lbs.Each$4,979.38: AC-250-3048
Specifications
Cat. No.89008-532 - Base cabinets, 2 framed acrylic sliding doorsPP42"13"30"135 lbs.Each$4,669.07: AC-250-3630
Specifications
Cat. No.89008-534 - Base cabinets, 2 framed acrylic sliding doorsPP48"13"30"160 lbs.Each$4,796.75: AC-250-4230
Specifications
Cat. No.89008-536 - Base cabinets, 2 framed acrylic sliding doorsPP30"13"24"185 lbs.Each$5,052.42: AC-250-4830
Specifications
Cat. No.89008-538 - Base cabinets, 2 framed acrylic sliding doorsPP36"13"24"90 lbs.Each$4,669.07: AC-250-3024
Specifications
Cat. No.89008-540 - Base cabinets, 2 framed acrylic sliding doorsPP42"13"24"110 lbs.Each$4,669.07: AC-250-3624
Specifications
Cat. No.89008-542 - Base cabinets, 2 framed acrylic sliding doorsPP48"13"24"130 lbs.Each$4,924.62: AC-250-4224
Specifications
Cat. No.89008-544 - Base cabinets, 2 framed acrylic sliding doorsPP18"13"30"150 lbs.Each$4,924.66: AC-250-4824
Specifications
Cat. No.89008-546 - : AC-205-3030
Specifications
Cat. No.89008-548 - : AC-205-3630
Specifications
Cat. No.89008-550 - : AC-205-4230
Specifications
Cat. No.89008-552 - : AC-205-4830
Specifications
Cat. No.89008-554 - : AC-205-3024
Specifications
Cat. No.89008-556 - : AC-205-3624
Specifications
Cat. No.89008-558 - : AC-205-4224
Specifications
Cat. No.89008-560 - : AC-205-4824
Specifications
Cat. No.89008-562 - Base cabinets, 1 framed acrylic hinged doorPP24"13"30"70 lbs.Each$4,290.26: AC-230-1830
Specifications
Cat. No.89008-564 - Base cabinets, 1 framed acrylic hinged doorPP30"13"30"85 lbs.Each$4,417.97: AC-230-2430
Specifications
Cat. No.89008-566 - Base cabinets, 1 framed acrylic hinged doorPP36"13"30"110 lbs.Each$4,545.80: AC-230-3030
Specifications
Cat. No.89008-568 - Base cabinets, 1 framed acrylic hinged doorPP18"13"24"135 lbs.Each$4,669.07: AC-230-3630
Specifications
Cat. No.89008-570 - Base cabinets, 1 framed acrylic hinged doorPP24"13"24"60 lbs.Each$4,162.47: AC-230-1824
Specifications
Cat. No.89008-572 - Base cabinets, 1 framed acrylic hinged doorPP30"13"24"80 lbs.Each$4,290.26: AC-230-2424
Specifications
Cat. No.89008-574 - Base cabinets, 1 framed acrylic hinged doorPP36"13"24"90 lbs.Each$4,417.98: AC-230-3024
Specifications
Cat. No.89008-576 - Base cabinets, 1 framed acrylic hinged doorPP30"13"30"110 lbs.Each$4,545.77: AC-230-3624
Specifications
Cat. No.89008-578 - : AC-200-1830
Specifications
Cat. No.89008-580 - : AC-200-2430
Specifications
Cat. No.89008-582 - : AC-200-3030
Specifications
Cat. No.89008-584 - : AC-200-3630
Specifications
Cat. No.89008-586 - : AC-200-1824
Specifications
Cat. No.89008-588 - : AC-200-2424
Specifications
Cat. No.89008-590 - : AC-200-3024
Specifications
Cat. No.89008-592 - : AC-200-3624
Specifications
Cat. No.89008-594 - Base cabinets, 2 framed acrylic hinged doorsPP36"13"30"110 lbs.Each$5,499.69: AC-210-3030
Specifications
Cat. No.89008-596 - Base cabinets, 2 framed acrylic hinged doorsPP42"13"30"135 lbs.Each$5,659.49: AC-210-3630
Specifications
Cat. No.89008-598 - Base cabinets, 2 framed acrylic hinged doorsPP48"13"30"160 lbs.Each$5,819.15: AC-210-4230
Specifications
Cat. No.89008-600 - Base cabinets, 2 framed acrylic hinged doorsPP30"13"24"185 lbs.Each$5,339.89: AC-210-4830
Specifications
Cat. No.89008-602 - Base cabinets, 2 framed acrylic hinged doorsPP36"13"24"90 lbs.Each$5,020.48: AC-210-3024
Specifications
Cat. No.89008-604 - Base cabinets, 2 framed acrylic hinged doorsPP42"13"24"110 lbs.Each$5,339.89: AC-210-3624
Specifications
Cat. No.89008-606 - Base cabinets, 2 framed acrylic hinged doorsPP48"13"24"130 lbs.Each$5,659.49: AC-210-4224
Specifications
Cat. No.89008-608 - Base cabinets, 2 framed acrylic hinged doorsPP48"13"48"140 lbs.Each$5,608.74: AC-210-4824
Specifications
Cat. No.89008-610 - : AC-210-4848
Specifications
Cat. No.89008-612 - : AC-220-3030
Specifications
Cat. No.89008-614 - : AC-220-3630
Specifications
Cat. No.89008-616 - : AC-220-4230
Specifications
Cat. No.89008-618 - : AC-220-4830
Specifications
Cat. No.89008-620 - : AC-220-3036
Specifications
Cat. No.89008-622 - : AC-220-3636
Specifications
Cat. No.89008-624 - : AC-220-4236
Specifications
Cat. No.89008-626 - : AC-220-4836
Specifications
Cat. No.89008-628 - : AC-220-5430
Specifications
Cat. No.89008-630 - : AC-220-7236
Specifications
Cat. No.89008-632 - Base cabinets, 2 solid hinged doors, for blind cornersPP50"13"30"125 lbs.Each$5,303.39: AC-215-3030
Specifications
Cat. No.89008-746 - Base cabinets, 2 solid hinged doors, for blind cornersPP56"13"30"160 lbs.Each$5,431.24: AC-215-5030
Specifications
Cat. No.89008-748 - Base cabinets, 2 solid hinged doors, for blind cornersPP62"13"30"140 lbs.Each$5,559.05: AC-215-4230
Specifications
Cat. No.89008-750 - Base cabinets, 2 solid hinged doors, for blind cornersPP44"13"24"150 lbs.Each$5,691.44: AC-215-4830
Specifications
Cat. No.89008-752 - Base cabinets, 2 solid hinged doors, for blind cornersPP50"13"24"110 lbs.Each$5,431.24: AC-215-3024
Specifications
Cat. No.89008-754 - Base cabinets, 2 solid hinged doors, for blind cornersPP56"13"24"145 lbs.Each$5,559.05: AC-215-3624
Specifications
Cat. No.89008-756 - Base cabinets, 2 solid hinged doors, for blind cornersPP62"13"24"125 lbs.Each$5,691.44: AC-215-4224
Specifications
Cat. No.89008-758 - Base cabinets, 2 solid hinged doors, for blind cornersPP30"13"30"135 lbs.Each$5,819.15: AC-215-4824
Specifications
Cat. No.89008-760